missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-pentene, 97%

Product Code. 10284980 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10284980 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10284980 Supplier Thermo Scientific Acros Supplier No. 127420250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 691-37-2 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 691-37-2
CAS Min % 96.0
CAS Max % 100.0
Melting Point -155°C
Color Colorless
Density 0.6600g/mL
Boiling Point 53°C to 54°C
Flash Point -31°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C6H12
Linear Formula (CH3)2CHCH2CH=CH2
Refractive Index 1.3810 to 1.383
MDL Number MFCD00008949
Quantity 25 mL
Specific Gravity 0.66
Synonym 4-methyl-1-pentene, 1-pentene, 4-methyl, poly 4-methyl-1-pentene, unii-x10hrj2y7w, x10hrj2y7w, poly 4-methylpentene, isobutylethene, isohexene, 4-methylpentene-1, 1-pentene, 4-methyl-, homopolymer
InChI Key WSSSPWUEQFSQQG-UHFFFAOYSA-N
SMILES CC(C)CC=C
IUPAC Name 4-methylpent-1-ene
Molecular Weight (g/mol) 84.16
PubChem CID 12724
Formula Weight 84.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-Methyl-1-pentene, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methyl-1-pentene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.