missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,2,3-thiadiazole-5-carboxylic Acid, TRC

Product Code. 30479276
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30479276 100 mg 100mg
30472945 500 mg 500mg
30470928 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479276 Supplier Toronto Research Chemicals Supplier No. TRCM331338100MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18212-21-0
Molecular Formula C4H4N2O2S
Molecular Weight (g/mol) 144.15
Synonym 1,2,3-Thiadiazole-5-carboxylic acid, 4-methyl-, 4-Methyl-1,2,3-thiadiazole-5-carboxylic Acid, NK-F 001, SV 03
IUPAC Name 4-methylthiadiazole-5-carboxylic acid
SMILES Cc1nnsc1C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C4H4N2O2S/c1-2-3(4(7)8)9-6-5-2/h1H3,(H,7,8)
Formula Weight 143.9993
Chemical Name or Material 4-Methyl-1,2,3-thiadiazole-5-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.