missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,2,3-thiadiazole-5-carbonyl chloride, Thermo Scientific™

Product Code. 10245074
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10245074 25 g Amber Glass Bottle 25g
10130922 1 g Amber Glass Bottle 1g
10517654 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10245074 Supplier Thermo Scientific Maybridge Supplier No. CC02302EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 59944-65-9
Molecular Formula C4H3ClN2OS
Molecular Weight (g/mol) 162.59
MDL Number MFCD00173831
InChI Key SDNDOCTUXWLDIX-UHFFFAOYSA-N
Synonym 4-methyl-1,2,3-thiadiazole-5-carbonyl chloride, 4-methyl-1,2,3-thiadiazole-5-carbonylchloride, 1,2,3-thiadiazole-5-carbonylchloride, 4-methyl, 4-methyl-1,2,3 thiadiazole-5-carbonyl chloride, 4-methyl-1,2,3-thiadiazole-5-carboxylic acid chloride, 4-methyl-5-thiadiazolecarbonyl chloride, 5-chlorocarbonyl-4-methyl-1,2,3-thiadiazole, 1,2,3-thiadiazole-5-carbonyl chloride, 4-methyl-9ci
PubChem CID 2736982
SMILES CC1=C(SN=N1)C(Cl)=O

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 25 g
IUPAC Name 4-methylthiadiazole-5-carbonyl chloride
Formula Weight 162.6
Percent Purity 90%
Chemical Name or Material 4-Methyl-1, 2, 3-thiadiazole-5-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.