missing translation for 'onlineSavingsMsg'
Learn More

4'-Methyl(1,1'-biphenyl)-4-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10265612
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265612 1 g Amber Glass Bottle 1g
10360823 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10265612 Supplier Thermo Scientific Maybridge Supplier No. MO07034DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 101366-51-2
Molecular Formula C13H11ClO2S
Molecular Weight (g/mol) 266.739
MDL Number MFCD01631922
InChI Key DBYXACYHGMSPMQ-UHFFFAOYSA-N
Synonym 4'-methylbiphenyl-4-sulfonyl chloride, 4'-methyl 1,1'-biphenyl-4-sulfonyl chloride, 4-4-methylphenyl benzenesulfonyl chloride, 4-4-methylphenyl phenyl sulphonyl chloride, 4'-methyl-1,1'-biphenyl-4-sulfonyl chloride, 4-p-tolyl benzenesulfonyl chloride, acmc-20amef, 4-4-chlorosulphonyl phenyl toluene, 4-chlorosulphonyl-4'-methylbiphenyl
PubChem CID 2794758
IUPAC Name 4-(4-methylphenyl)benzenesulfonyl chloride
SMILES CC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 266.75
Percent Purity 97%
Chemical Name or Material 4'-Methyl[1, 1'-biphenyl]-4-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.