missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methoxytrityl chloride, 97%

Brand:  Alfa Aesar™ A10545.18

Code : NEW

Additional Details : CAS Number : 14470-28-1 Weight : 0.05000kg

Quantity 50g

 View more versions of this product

Product Code. 11478823

  • £84.00 / 50g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Description

Description

Applications
It is used as selective protecting reagent that is used for primary hydroxyl groups.

Solubility
Soluble in toluene.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

4-Methoxytrityl chloride
C20H17ClO
MFCD00000814
p-Anisylchlorodiphenylmethane; 4-Methoxytriphenylmethyl chloride
14470-28-1
308.81
798425

Specifications

4-Methoxytrityl chloride
97%
308.81
798425
50g
14470-28-1
C20H17ClO
MFCD00000814
121° to 125°C
p-Anisylchlorodiphenylmethane; 4-Methoxytriphenylmethyl chloride
Videos
SDS