missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxytriphenylmethylchloride, TRC

Product Code. 30465630
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30465630 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30465630 Supplier Toronto Research Chemicals Supplier No. TRCM271660100G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methoxyriphenylmethylchloride
CAS 14470-28-1
Molecular Formula C20H17ClO
Quantity 100 g
Synonym 1-(Chlorodiphenylmethyl)-4-methoxybenzene, p-(Chlorodiphenylmethyl)anisole, (p-Anisyl)diphenylmethyl Chloride, 1-(Chlorodiphenylmethyl)-4-methoxybenzene, 4-Anisyl(chloro)diphenylmethane, 4-Methoxytriphenylmethyl Chloride, 4-Methoxytritylchloride, 4-Monomethoxytrityl Chloride, MMTCl, Mono-p-methoxytrityl Chloride, NSC 54121, p-(Chlorodiphenylmethyl)anisole, p-Anisylchlorodiphenylmethane, p-Methoxytrityl Chloride
InChI Formula InChI=1S/C20H17ClO/c1-22-19-14-12-18(13-15-19)20(21,16-8-4-2-5-9-16)17-10-6-3-7-11-17/h2-15H,1H3
SMILES COc1ccc(cc1)C(Cl)(c2ccccc2)c3ccccc3
IUPAC Name 1-[chloro(diphenyl)methyl]-4-methoxybenzene
Molecular Weight (g/mol) 308.8
Formula Weight 308.0968
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.