missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxypropiophenone, 99%

Product Code. 10396620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10396620 100 g Glass bottle 100g
10177613 5 g Glass bottle 5g
10674982 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10396620 Supplier Thermo Scientific Acros Supplier No. 126091000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 121-97-1 | C10H12O2 | 164.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-97-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00009310
InChI Key ZJVAWPKTWVFKHG-UHFFFAOYSA-N
PubChem CID 67144
IUPAC Name 1-(4-methoxyphenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 27°C to 29°C
Color Amber to Colorless
Density 1.0700g/mL
Boiling Point 273°C to 275°C
Flash Point 61°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3OC6H4COC2H5
Refractive Index 1.5455 to 1.5475
Quantity 100 g
Beilstein 08, 103
Specific Gravity 1.07
Synonym 4'-methoxypropiophenone, p-methoxypropiophenone, 4-methoxypropiophenone, 1-4-methoxyphenyl propan-1-one, 1-propanone, 1-4-methoxyphenyl, propiophenone, 4'-methoxy, ethyl 4-methoxyphenyl ketone, 1-4-methoxyphenyl-1-propanone, 1-propanone,1-4-methoxyphenyl, 1-4-methoxy-phenyl-propan-1-one
Formula Weight 164.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4'-Methoxypropiophenone, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Methoxypropiophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.