missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxypiperidine, 98+%

Product Code. 10248753 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10248753

Brand: Thermo Scientific Acros 393390050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4045-24-3 | C6H13NO | 115.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4045-24-3
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.18
MDL Number MFCD04115010
InChI Key ZEYSHALLPAKUHG-UHFFFAOYSA-N
Synonym 4-methoxy-piperidine, piperidine, 4-methoxy, hcl, 4-methoxypiperdine, 4-methoxy piperidine, 4-methoxyl piperidine, pubchem8024
PubChem CID 77666
IUPAC Name 4-methoxypiperidine
SMILES COC1CCNCC1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Density 0.9300g/mL
Boiling Point 44.0°C (9.0 Mbar)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4590 to 1.4610 (20°C, 589nm)
Quantity 5 g
Beilstein 21, V,1, 51
Specific Gravity 0.93
Formula Weight 115.18
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 4-Methoxypiperidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Methoxypiperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.