missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylhydrazine hydrochloride, 98%

Product Code. 11362356
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362356 5 g 5g
11372356 25 g 25g
11382356 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11362356 Supplier Thermo Scientific Alfa Aesar Supplier No. L06076.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 19501-58-7 | C7H11ClN2O | 174.628 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19501-58-7
Molecular Formula C7H11ClN2O
Molecular Weight (g/mol) 174.628
MDL Number MFCD00012945
InChI Key FQHCPFMTXFJZJS-UHFFFAOYSA-N
Synonym 4-methoxyphenylhydrazine hydrochloride, 4-methoxyphenyl hydrazine hydrochloride, 4-anisylhydrazine hydrochloride, 4-methoxyphenylhydrazine hcl, p-methoxyphenylhydrazine hydrochloride, 4-methoxyphenylhydrazine.hcl, 1-4-methoxyphenyl hydrazine hydrochloride, p-anisylhydrazine hydrochloride, 4-hydrazinoanisole hydrochloride, hydrazine, 4-methoxyphenyl-, monohydrochloride
PubChem CID 2723904
IUPAC Name (4-methoxyphenyl)hydrazine;hydrochloride
SMILES COC1=CC=C(C=C1)NN.Cl

Specifications

Melting Point ∽161°C (decomposition)
Quantity 5 g
Beilstein 3566583
Formula Weight 174.63
Percent Purity 99%
Chemical Name or Material 4-Methoxyphenylhydrazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.