missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylhydrazine hydrochloride, 98%

Product Code. p-158665 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10321384 1 g Glass bottle 1g
10275640 10 g Glass bottle 10g
10732591 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10321384 Supplier Thermo Scientific Acros Supplier No. 166490010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 19501-58-7 | C7H10N2O·ClH | 174.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19501-58-7
Molecular Formula C7H10N2O·ClH
Molecular Weight (g/mol) 174.63
MDL Number MFCD00012945
InChI Key FQHCPFMTXFJZJS-UHFFFAOYSA-N
Synonym 4-methoxyphenylhydrazine hydrochloride, 4-methoxyphenyl hydrazine hydrochloride, 4-anisylhydrazine hydrochloride, 4-methoxyphenylhydrazine hcl, p-methoxyphenylhydrazine hydrochloride, 4-methoxyphenylhydrazine.hcl, 1-4-methoxyphenyl hydrazine hydrochloride, p-anisylhydrazine hydrochloride, 4-hydrazinoanisole hydrochloride, hydrazine, 4-methoxyphenyl-, monohydrochloride
PubChem CID 2723904
IUPAC Name (4-methoxyphenyl)hydrazine;hydrochloride
SMILES COC1=CC=C(C=C1)NN.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 153°C to 154°C
Color Pink to Purple
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3OC6H4NHNH2·HCl
Quantity 1 g
Beilstein 15, 597
Solubility Information Solubility in water: soluble
Formula Weight 174.63
Percent Purity 98%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 4-Methoxyphenylhydrazine hydrochloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.