missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylglyoxal Hydrate, TRC

Product Code. 30478197
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478197 100 mg 100mg
30464711 500 mg 500mg
30472658 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478197 Supplier Toronto Research Chemicals Supplier No. TRCM220475100MG
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methoxyphenylglyoxal Hydrate
CAS 16208-17-6
Molecular Formula C9 H10 O4
Quantity 100 mg
InChI Formula InChI=1S/C9H10O4/c1-13-7-4-2-6(3-5-7)8(10)9(11)12/h2-5,9,11-12H,1H3
SMILES COc1ccc(cc1)C(=O)C(O)O
IUPAC Name 2,2-dihydroxy-1-(4-methoxyphenyl)ethanone
Molecular Weight (g/mol) 182.1733
Formula Weight 182.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.