missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylglyoxal hydrate, 95%, dry wt. basis

Product Code. 11399136
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399136 1 g 1g
11309146 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399136 Supplier Thermo Scientific Alfa Aesar Supplier No. L20274.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16208-17-6 | C9H10O4 | 182.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16208-17-6
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00234534
InChI Key NLUFPYPVEHQAQN-UHFFFAOYSA-N
Synonym 4-methoxyphenylglyoxal hydrate, 2-4-methoxyphenyl-2-oxoacetaldehyde hydrate, pubchem12129, acmc-20ai17, 4-methoxyphenyl glyoxal hydrate, p-methoxyphenylglyoxal monohydrate, 2-4-methoxyphenyl-2-oxoethanal, hydrate, ethanone,2,2-dihydroxy-1-4-methoxyphenyl, 4-methoxyphenyl oxo acetaldehyde-water 1/1, 2-4-methoxyphenyl-2-oxidanylidene-ethanal hydrate
PubChem CID 15556730
IUPAC Name 2-(4-methoxyphenyl)-2-oxoacetaldehyde;hydrate
SMILES COC1=CC=C(C=C1)C(=O)C(O)O

Specifications

Melting Point 104°C to 111°C
Boiling Point 145°C to 147°C (13 mmHg)
Quantity 1 g
Beilstein 606701
Formula Weight 164.16 (Anhydrous)
Percent Purity 95%
Assay (dry wt.)
Chemical Name or Material 4-Methoxyphenylglyoxal hydrate
compliance-icons
Product Identifier
  • 4-Methoxyphenylglyoxal hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Methoxyphenylglyoxal hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.