missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number 10328790
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5720-07-0
C7H9BO3
151.96
MFCD00039139
VOAAEKKFGLPLLU-UHFFFAOYSA-N
4-methoxyphenyl boronic acid, 4-methoxybenzeneboronic acid, p-anisylboronic acid, p-methoxyphenylboronic acid, p-methoxybenzeneboronic acid, 4-methoxyphenyl boranediol, benzeneboronic acid, p-methoxy, 4-boronoanisole, 4-methoxyphenylboronicacid
201262
(4-methoxyphenyl)boronic acid
COC1=CC=C(C=C1)B(O)O
This item is not returnable. View return policy
5720-07-0
C7H9BO3
151.96
MFCD00039139
VOAAEKKFGLPLLU-UHFFFAOYSA-N
4-methoxyphenyl boronic acid, 4-methoxybenzeneboronic acid, p-anisylboronic acid, p-methoxyphenylboronic acid, p-methoxybenzeneboronic acid, 4-methoxyphenyl boranediol, benzeneboronic acid, p-methoxy, 4-boronoanisole, 4-methoxyphenylboronicacid
201262
(4-methoxyphenyl)boronic acid
COC1=CC=C(C=C1)B(O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 208.0°C to 210.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OC6H4B(OH)2
Quantity 1 g
Formula Weight 151.96
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenylboronic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'