missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylacetylene, 98%

Product Code. 11366779
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11366779 5 g 5g
11356779 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11366779 Supplier Thermo Scientific Alfa Aesar Supplier No. H26283.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 768-60-5 | C9H8O | 132.162 g/mol

4-Ethynylanisole was used in the synthesis of photo luminescent 1,2-dihydrophosphinines via a [4 + 2] cycloaddition. Along with an arylboronic acid and sodium azide in a copper-catalyzed, three-component synthesis of trisubstituted 1,2,4-triazoles. In a study of a gold (III)-catalyzed hydroamination of alkynes leading to N-vinylindoles3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethynylanisole was used in the synthesis of photo luminescent 1,2-dihydrophosphinines via a [4 + 2] cycloaddition. Along with an arylboronic acid and sodium azide in a copper-catalyzed, three-component synthesis of trisubstituted 1,2,4-triazoles. In a study of a gold (III)-catalyzed hydroamination of alkynes leading to N-vinylindoles3.

Solubility
Insoluble in water. Soluble in chloroform, acetone, dichloromethane, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-60-5
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
MDL Number MFCD00168815
InChI Key KBIAVTUACPKPFJ-UHFFFAOYSA-N
Synonym 4-ethynylanisole, 4-methoxyphenylacetylene, 4'-methoxyphenyl acetylene, 1-ethynyl-4-methoxy-benzene, 1-eth-1-ynyl-4-methoxybenzene, benzene, 1-ethynyl-4-methoxy, 4-methoxyphenyl acetylene, 4-ethynyl-1-methoxybenzene, 4'-methoxyphenylacetylene, p-ethynylanisole
PubChem CID 251020
IUPAC Name 1-ethynyl-4-methoxybenzene
SMILES COC1=CC=C(C=C1)C#C

Specifications

Melting Point 27°C to 29°C
Density 1.019
Boiling Point 87°C to 91°C (11 mmHg)
Flash Point 82°C (180°F)
Refractive Index 1.563
Quantity 5 g
Solubility Information Insoluble in water. Soluble in chloroform,acetone,dichloromethane,and methanol.
Formula Weight 132.16
Percent Purity 99%
Chemical Name or Material 4-Methoxyphenylacetylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.