missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylacetone, 97+%

Product Code. 11450723
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450723 500 g 500g
11434644 25 g 25g
11444644 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450723 Supplier Thermo Scientific Alfa Aesar Supplier No. A13209.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-84-9 | C10H12O2 | 164.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-84-9
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00008773
InChI Key WFWKNGZODAOLEO-UHFFFAOYSA-N
Synonym 4-methoxyphenylacetone, p-acetonylanisole, anisketone, 1-4-methoxyphenyl propan-2-one, 2-propanone, 1-4-methoxyphenyl, anisyl methyl ketone, p-methoxyphenylacetone, 4-methoxybenzyl methyl ketone, 1-p-methoxyphenyl-2-propanone, anisic ketone
PubChem CID 31231
IUPAC Name 1-(4-methoxyphenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(C=C1)OC

Specifications

Density 1.069
Boiling Point 266°C to 268°C
Flash Point 101°C (213°F)
Refractive Index 1.525
Quantity 500 g
Beilstein 2044332
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material 4-Methoxyphenylacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.