missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylacetic acid, 99%

Product Code. 10424585 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
1kg
250g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10424585 1 kg Plastic bottle 1kg
10347010 5 g Glass bottle 5g
10785041 100 g Plastic bottle 100g
10307060 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10424585 Supplier Thermo Scientific Acros Supplier No. 126020010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-01-8 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-01-8
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00004345
InChI Key NRPFNQUDKRYCNX-UHFFFAOYSA-N
Synonym 4-methoxyphenylacetic acid, homoanisic acid, 2-4-methoxyphenyl acetic acid, 4-methoxybenzeneacetic acid, benzeneacetic acid, 4-methoxy, 4-methoxyphenyl acetic acid, p-methoxyphenylacetic acid, 2-p-anisyl acetic acid, p-methoxyphenyl acetic acid
PubChem CID 7690
ChEBI CHEBI:55501
IUPAC Name 2-(4-methoxyphenyl)acetic acid
SMILES COC1=CC=C(C=C1)CC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 84.0°C to 88.0°C
Color White to Yellow
Boiling Point 140.0°C (3.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula CH3OC6H4CH2CO2H
Quantity 1 kg
Beilstein 10, 190
Solubility Information Solubility in water: 6g/L (20 c)
Formula Weight 166.18
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4-Methoxyphenylacetic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxyphenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.