missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, is mainly used as a nematic liquid crystal monomer, liquid crystal material. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Sparingly Soluble in water (1.7E-3 g/L) (25°C).
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate, is mainly used as a nematic liquid crystal monomer, liquid crystal material. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Sparingly Soluble in water (1.7E-3 g/L) (25°C).
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Melting Point | 40°C to 44°C |
Quantity | 1 g |
Solubility Information | Sparingly Soluble in water (1.7E-3g/L) (25°C). |
Formula Weight | 304.43 |
Percent Purity | 97% |
Chemical Name or Material | 4-Methoxyphenyl trans-4-n-pentylcyclohexanecarboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'