missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl isothiocyanate, 98%

Product Code. 11458236
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458236 50 g 50g
11394699 2 g 2g
11448236 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458236 Supplier Thermo Scientific Alfa Aesar Supplier No. A15331.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2284-20-0 | C8H7NOS | 165.21 g/mol

4-Methoxyphenyl isothiocyanate has been used in the synthesis of 3-benzyl-2-(4-methoxyphenyl)-3H-[1,2,4]triazolo-[5,1-b]quinazolin-9-one and 2-{[4-amino-3-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetyl}-N-arylhydrazinecarbothioamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxyphenyl isothiocyanate has been used in the synthesis of 3-benzyl-2-(4-methoxyphenyl)-3H-[1,2,4]triazolo-[5,1-b]quinazolin-9-one and 2-{[4-amino-3-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetyl}-N-arylhydrazinecarbothioamides.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2284-20-0
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00011676
InChI Key VRPQCVLBOZOYCG-UHFFFAOYSA-N
Synonym 4-methoxyphenyl isothiocyanate, 4-methoxyphenylisothiocyanate, 1-isothiocyanato-4-methoxy-benzene, p-methoxyphenyl isothiocyanate, benzene, 1-isothiocyanato-4-methoxy, isothiocyanic acid 4-methoxyphenyl ester, 4-methoxybenzenisothiocyanate, 4-isothiocyanatoanisole, acmc-1ccqx, p-methoxyphenylisothiocyanate
PubChem CID 75293
IUPAC Name 1-isothiocyanato-4-methoxybenzene
SMILES COC1=CC=C(C=C1)N=C=S

Specifications

Melting Point 18°C to 20°C
Density 1.196
Boiling Point 279°C to 281°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.6489
Quantity 50 g
UN Number UN2922
Beilstein 606967
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes in water.
Formula Weight 165.22
Percent Purity 98%
Chemical Name or Material 4-Methoxyphenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.