missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxyphenyl chloroformate, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
Specifications
Specifications
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Color | Colorless to Yellow |
| Density | 1.25g/mL |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass Bottle |
| Linear Formula | ClCOC6H4OCH3 |
| Refractive Index | 1.5220 to 1.5260 (20°C, 589nm) |
| Quantity | 25 g |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction