missing translation for 'onlineSavingsMsg'
Learn More

(4-Methoxyphenyl)acetonitrile, 97%

Product Code. 10317310 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10317310 25 g Glass bottle 25g
10343382 5 g Glass bottle 5g
10301574 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10317310 Supplier Thermo Scientific Acros Supplier No. 173760250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-47-2 | C9H9NO | 147.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-47-2
MDL Number MFCD00001919
InChI Key PACGLQCRGWFBJH-UHFFFAOYSA-N
Synonym 4-methoxyphenylacetonitrile, 2-4-methoxyphenyl acetonitrile, 4-methoxybenzyl cyanide, 4-methoxyphenyl acetonitrile, p-methoxyphenylacetonitrile, benzeneacetonitrile, 4-methoxy, p-methoxybenzyl cyanide, anisylacetonitrile, 4-anisyl cyanide, p-methoxybenzeneacetonitrile
PubChem CID 66031
IUPAC Name 2-(4-methoxyphenyl)acetonitrile
SMILES COC1=CC=C(C=C1)CC#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 8.0°C
Color Colorless to Yellow
Density 1.0800g/mL
Boiling Point 286.0°C to 287.0°C
Flash Point 117°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H9NO
Linear Formula CH3OC6H4CH2CN
Refractive Index 1.53
Quantity 25 g
Beilstein 10, 191
Specific Gravity 1.08
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in ethyl alcohol,ethyl ether,chloroform,miscible in isopropanol,acetone,toluene
Molecular Weight (g/mol) 147.18
Formula Weight 147.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (4-Methoxyphenyl)acetonitrile
Show More Show Less
compliance-icons
Product Identifier
  • (4-Methoxyphenyl)acetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.