missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenoxyacetonitrile, 97%, Thermo Scientific™

Product Code. 11448583
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448583 5 g 5g
11458583 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448583 Supplier Thermo Scientific Alfa Aesar Supplier No. A10398.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It finds it application in the synthesis of α-aryloxy-2,4-dihydroxyacetophenone 4-O-β-D-glucopyranosides and their conversion into 3-aryloxy-7-glucosyloxychromones.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22446-12-4
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00017528
InChI Key SNADCCRKYFVIEJ-UHFFFAOYSA-N
Synonym 4-methoxyphenoxyacetonitrile, 2-4-methoxyphenoxy acetonitrile, 4-methoxyphenoxy acetonitrile, acetonitrile, 4-methoxyphenoxy, 2-4-methoxyphenoxy ethanenitrile, acmc-20amep, p-methoxy-phenoxyacetonitrile, 4-methoxyphenoxy acetonitrile #, 2-4-methoxyphenoxy-acetonitrile, acetonitrile,2-4-methoxyphenoxy
PubChem CID 579491
IUPAC Name 2-(4-methoxyphenoxy)acetonitrile
SMILES COC1=CC=C(C=C1)OCC#N

Specifications

Boiling Point 106°C to 110°C (0.1 mmHg)
Refractive Index 1.527
Quantity 5 g
UN Number UN3276
Beilstein 2444937
Formula Weight 163.18
Percent Purity 97%
Chemical Name or Material 4-Methoxyphenoxyacetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.