missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenethyl alcohol, 96%

Product Code. 10103391 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10103391 50 g Glass bottle 50g
10327751 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10103391 Supplier Thermo Scientific Acros Supplier No. 354050500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 702-23-8 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 702-23-8
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002900
InChI Key AUWDOZOUJWEPBA-UHFFFAOYSA-N
Synonym 2-4-methoxyphenyl ethanol, 4-methoxyphenethyl alcohol, 4-methoxybenzeneethanol, benzeneethanol, 4-methoxy, p-methoxyphenethyl alcohol, 2-4-methoxyphenyl ethan-1-ol, unii-3mwl5nze9k, 3mwl5nze9k, 4-2-hydroxyethyl anisole, 2-4-methoxyphenyl-ethanol
PubChem CID 69705
IUPAC Name 2-(4-methoxyphenyl)ethanol
SMILES COC1=CC=C(C=C1)CCO

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 28°C to 30°C
Color Brown
Boiling Point 334°C to 336°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4CH2CH2OH
Refractive Index 1.5360 to 1.538
Quantity 50 g
Beilstein 06, 906
Formula Weight 152.19
Percent Purity 96%
Physical Form Liquid After Melting
Chemical Name or Material 4-Methoxyphenethyl alcohol, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.