missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxynaphthalene-1-carboxylic acid, 98%

Product Code. p-7055769
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15441639

Brand: Thermo Scientific Alfa Aesar H64432.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13041-62-8 | C12H10O3 | 202.209 g/mol

Used in the preparation of α-aryl/pyridinyl ethanolamines as selective β3 adrenergic receptor agonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of α-aryl/pyridinyl ethanolamines as selective β3 adrenergic receptor agonists.

Solubility
Soluble in DMSO and chloroform. Slightly soluble in water.

Notes
Store away from acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13041-62-8
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.209
MDL Number MFCD02575372
InChI Key WRQHSQDGYYDRMX-UHFFFAOYSA-N
Synonym 4-methoxy-1-naphthoic acid, 4-methoxy-naphthalene-1-carboxylic acid, 1-naphthalenecarboxylic acid, 4-methoxy, 1-naphthalenecarboxylicacid, 4-methoxy, 4-methoxynaphthalenecarboxylic acid, ksc181m4d, 4-methoxy-1-naphthalenecarboxylic acid
PubChem CID 765380
IUPAC Name 4-methoxynaphthalene-1-carboxylic acid
SMILES COC1=CC=C(C2=CC=CC=C21)C(=O)O

Specifications

Melting Point 243°C to 247°C
Color White
Quantity 1 g
Beilstein 2099800
Solubility Information Soluble in DMSO and chloroform. Slightly soluble in water.
Formula Weight 202.21
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 4-Methoxynaphthalene-1-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.