missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxymethyl)benzeneboronic acid, 97%

Product Code. 15488368
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15488368 25 g 25g
15478368 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488368 Supplier Thermo Scientific Alfa Aesar Supplier No. H61795.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 279262-11-2 | C8H11BO3 | 165.983 g/mol

4-(Methoxymethyl)benzeneboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methoxymethyl)benzeneboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 279262-11-2
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.983
MDL Number MFCD03788426
InChI Key VTNVZXAYRMTKON-UHFFFAOYSA-N
Synonym 4-methoxymethyl phenylboronic acid, 4-methoxymethyl phenyl boronic acid, 4-methoxymethylbenzeneboronic acid, 4-methoxymethylphenylboronic acid, 4-methoxymethyl benzeneboronic acid, acmc-1cr68, 4-methoxymethyl-phenylboronic acid, 4-methoxymethylphenyl boronic acid, 4-methoxymethyl-benzene boronic acid
PubChem CID 3723812
IUPAC Name [4-(methoxymethyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)COC)(O)O

Specifications

Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 165.99
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(Methoxymethyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.