missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxymetanilyl fluoride, 97%, Thermo Scientific™

Product Code. 10173620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173620 25 g Glass bottle 25g
10113580 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10173620 Supplier Thermo Scientific Acros Supplier No. 148330250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-74-8
Molecular Formula C7H8FNO3S
Molecular Weight (g/mol) 205.2
MDL Number MFCD00007417
InChI Key SESLQGPHIQXYGF-UHFFFAOYSA-N
Synonym 4-methoxymetanilyl fluoride, 3-amino-4-methoxyphenylsulphonyl fluoride, 3-amino-4-methoxy benzene sulphonylfluoride, 3-amino-4-methoxyphenyl fluorosulfone, 3-amino-4-methoxy-benzenesulfonyl fluoride, 3-amino-4-methoxybenzenesulphonyl fluoride, 3-azanyl-4-methoxy-benzenesulfonyl fluoride, benzenesulfonylfluoride, 3-amino-4-methoxy, 3-amino-4-methoxybenzene-1-sulfonyl fluoride, benzenesulfonyl fluoride, 3-amino-4-methoxy
PubChem CID 96952
IUPAC Name 3-amino-4-methoxybenzenesulfonyl fluoride
SMILES COC1=C(C=C(C=C1)S(=O)(=O)F)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 60°C to 65°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OC6H3(NH2)SO2F
Quantity 25 g
Beilstein 14, II, 487
Formula Weight 205.2
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 4-Methoxymetanilyl fluoride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxymetanilyl fluoride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.