missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxymetanilyl fluoride, 97%, Thermo Scientific™

Product Code. 10113580
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-74-8
Molecular Formula C7H8FNO3S
Molecular Weight (g/mol) 205.2
MDL Number MFCD00007417
InChI Key SESLQGPHIQXYGF-UHFFFAOYSA-N
Synonym 4-methoxymetanilyl fluoride, 3-amino-4-methoxyphenylsulphonyl fluoride, 3-amino-4-methoxy benzene sulphonylfluoride, 3-amino-4-methoxyphenyl fluorosulfone, 3-amino-4-methoxy-benzenesulfonyl fluoride, 3-amino-4-methoxybenzenesulphonyl fluoride, 3-azanyl-4-methoxy-benzenesulfonyl fluoride, benzenesulfonylfluoride, 3-amino-4-methoxy, 3-amino-4-methoxybenzene-1-sulfonyl fluoride, benzenesulfonyl fluoride, 3-amino-4-methoxy
PubChem CID 96952
IUPAC Name 3-amino-4-methoxybenzenesulfonyl fluoride
SMILES COC1=C(C=C(C=C1)S(=O)(=O)F)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 60°C to 65°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OC6H3(NH2)SO2F
Quantity 10 g
Beilstein 14, II, 487
Formula Weight 205.2
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 4-Methoxymetanilyl fluoride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxymetanilyl fluoride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'