missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxycarbonyl)phenyl isothiocyanate, 98+%

Product Code. 11398445
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398445 5 g 5g
11388445 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398445 Supplier Thermo Scientific Alfa Aesar Supplier No. L10474.06
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3662-78-0 | C9H7NO2S | 193.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3662-78-0
Molecular Formula C9H7NO2S
Molecular Weight (g/mol) 193.22
MDL Number MFCD00041099
InChI Key WIZODHILGBTPPA-UHFFFAOYSA-N
Synonym 4-methoxycarbonylphenyl isothiocyanate, 4-methoxycarbonylphenylisothiocyanate, 4-isothiocyanatobenzoic acid methyl ester, benzoic acid, 4-isothiocyanato-, methyl ester, acmc-20amez, #, 4-methoxycarbonyl phenylisothiocyanate, 4-methoxycarbonyl phenyl isothiocyanate
PubChem CID 138002
IUPAC Name methyl 4-isothiocyanatobenzoate
SMILES COC(=O)C1=CC=C(C=C1)N=C=S

Specifications

Melting Point 46°C to 50°C
Odor Irritating
Quantity 5 g
UN Number UN2923
Beilstein 2100504
Sensitivity Moisture sensitive
Formula Weight 193.23
Percent Purity ≥98%
Chemical Name or Material 4-(Methoxycarbonyl)phenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.