missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxycarbonyl)cyclohexane-1-carboxylic acid, 97%

Product Code. 15440136
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15440136 5 g 5g
15430136 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440136 Supplier Thermo Scientific Alfa Aesar Supplier No. H52814.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 32529-79-6 | C9H14O4 | 186.207 g/mol

4-(Methoxycarbonyl)cyclohexane-1-carboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methoxycarbonyl)cyclohexane-1-carboxylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water (11 g/L at 25°C).

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32529-79-6
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.207
MDL Number MFCD06797561
InChI Key ZQJNPHCQABYENK-UHFFFAOYSA-N
Synonym trans-1,4-cyclohexanedicarboxylic acid monomethyl ester, cis-4-methoxycarbonyl cyclohexanecarboxylic acid, trans-4-methoxycarbonyl cyclohexanecarboxylic acid, 4-methoxycarbonyl cyclohexanecarboxylic acid, 1r,4r-4-methoxycarbonyl cyclohexanecarboxylic acid, 4-methoxycarbonyl cyclohexane-1-carboxylic acid, 4-carbomethoxy-cyclohexane-1-carboxylic acid, trans-4-carbomethoxycyclohexane-1-carboxylic acid, 1r,4r-4-methoxycarbonyl cyclohexane-1-carboxylic acid, trans-1,4-cyclohexane dicarboxylic acid monomethyl ester
PubChem CID 296850
IUPAC Name 4-methoxycarbonylcyclohexane-1-carboxylic acid
SMILES COC(=O)C1CCC(CC1)C(=O)O

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water (11g/L at 25°C).
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 4-(Methoxycarbonyl)cyclohexane-1-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.