missing translation for 'onlineSavingsMsg'
Learn More

(4-Methoxybenzyl)triphenylphosphonium chloride, 98%, Thermo Scientific™

Product Code. 10340374 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10340374 5 g Glass Bottle 5g
10704004 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10340374 Supplier Thermo Scientific Acros Supplier No. 376320050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3462-97-3
Molecular Formula C26H24ClOP
Molecular Weight (g/mol) 418.90
MDL Number MFCD00031572
InChI Key YQXBNCFNXOFWLR-UHFFFAOYSA-M
Synonym 4-methoxybenzyl triphenylphosphonium chloride, 4-methoxyphenyl methyl triphenylphosphanium chloride, acmc-1ctv8, 4-methoxyphenyl methyl-triphenyl-phosphanium chloride, 4-methoxybenzyltriphenylphosphonium chloride, 4-methoxybenzyltriphenylphosphonium-chloride, 4-methoxybenzyl triphenyphosphonium chloride, p-methoxybenzyl-triphenylphosphonium chloride, 4-methoxybenzyltriphenyl phosphonium chloride, 4-methoxybenzyl triphenyl phosphonium chloride
PubChem CID 2773651
SMILES [Cl-].COC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 244.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CH3OC6H4CH2P(C6H5)3Cl
Quantity 5 g
IUPAC Name [(4-methoxyphenyl)methyl]triphenylphosphanium chloride
Formula Weight 418.9
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (4-Methoxybenzyl)triphenylphosphonium chloride
Show More Show Less
compliance-icons
Product Identifier
  • (4-Methoxybenzyl)triphenylphosphonium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.