missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl mercaptan, 98%

Product Code. 11445107
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445107 5 g 5g
11455107 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445107 Supplier Thermo Scientific Alfa Aesar Supplier No. B22542.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6258-60-2 | C8H10OS | 154.23 g/mol

4-Methoxybenzyl mercaptan is used to synthesize many ω-mercapto amino acids with side-chain lengths ranging from 3-5 methylene units. It is used as an internal standard in the Quantitative determination of wine polyfunctional mercaptans at the level of nanogram per liter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzyl mercaptan is used to synthesize many ω-mercapto amino acids with side-chain lengths ranging from 3-5 methylene units. It is used as an internal standard in the Quantitative determination of wine polyfunctional mercaptans at the level of nanogram per liter.

Solubility
Not miscible or difficult to mix in water.

Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agents, acids, bases, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6258-60-2
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.23
MDL Number MFCD00004871
InChI Key PTDVPWWJRCOIIO-UHFFFAOYSA-N
Synonym 4-methoxybenzyl mercaptan, 4-methoxy-alpha-toluenethiol, 4-methoxybenzylmercaptan, 4-methoxyphenyl methanethiol, 4-methoxy benzylmercaptan, 4-methoxybenzenemethanethiol, benzenemethanethiol, 4-methoxy, p-methoxybenzyl mercaptan, p-methoxybenzylthiol, p-methoxybenzylmercaptan
PubChem CID 80407
IUPAC Name (4-methoxyphenyl)methanethiol
SMILES COC1=CC=C(CS)C=C1

Specifications

Density 1.112
Boiling Point 90°C to 95°C (0.5 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.5755
Quantity 5 g
Beilstein 471686
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 154.23
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzyl mercaptan
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.