missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl Alcohol, TRC

Product Code. 30471051
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471051 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471051 Supplier Toronto Research Chemicals Supplier No. TRCM2610655G
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methoxybenzyl alcohol (Anisyl alcohol)
CAS 105-13-5
Purity Grade Notes HPLC
Molecular Formula C8 H10 O2
Quantity 5 g
Synonym Benzyl alcohol, p-methoxy- (8CI), 4-Methoxybenzenemethanol, (4-Methoxyphenyl)methyl alcohol, 4-(Hydroxymethyl)anisole, 4-(Methoxyphenyl)methanol, 4-Methoxy-α-hydroxytoluene, 4-Methoxybenzyl alcohol, Anise alcohol, Anisic alcohol, NSC 2151, [4-(Methyloxy)phenyl]methanol, p-(Methoxyphenyl)methanol, p-Anisalcohol, p-Anisyl alcohol, p-Methoxybenzyl alcohol
InChI Formula InChI=1S/C8H10O2/c1-10-8-4-2-7(6-9)3-5-8/h2-5,9H,6H2,1H3
SMILES COc1ccc(CO)cc1
IUPAC Name (4-methoxyphenyl)methanol
Molecular Weight (g/mol) 138.16
Formula Weight 138.0681
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.