missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzoyl Chloride, TRC

Product Code. 30466693
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466693 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466693 Supplier Toronto Research Chemicals Supplier No. TRCM28891550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methoxybenzoyl Chloride
CAS 100-07-2
Molecular Formula C8H7ClO2
Quantity 50 g
Synonym p-Anisoyl Chloride, 4-Anisoyl Chloride, 4-Methoxybenzoic Acid Chloride, 4-Methoxybenzoyl Chloride, 4-Methoxyphenylchloroform, Anisoyl Chloride, NSC 86125, Para-anisoyl Chloride, p-Methoxybenzoic Acid Chloride, p-Methoxybenzoyl Chloride
InChI Formula InChI=1S/C8H7ClO2/c1-11-7-4-2-6(3-5-7)8(9)10/h2-5H,1H3
SMILES COc1ccc(cc1)C(=O)Cl
Molecular Weight (g/mol) 170.59
Formula Weight 170.0135
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.