missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzoic anhydride, 98%

Product Code. p-7058306
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11322406

Brand: Thermo Scientific Alfa Aesar L06180.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 794-94-5 | C16H14O5 | 286.283 g/mol

4-Methoxybenzoic anhydride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzoic anhydride is used as pharmaceutical intermediates.

Solubility
Soluble in alcohol, ether. Insoluble in water.

Notes
Moisture Sensitive. Protect from humidity and water. Incompatible with moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 794-94-5
Molecular Formula C16H14O5
Molecular Weight (g/mol) 286.283
MDL Number MFCD00017175
InChI Key YGMHIBLUWGDWKP-UHFFFAOYSA-N
Synonym 4-methoxybenzoic anhydride, p-anisic anhydride, p-anisic acid anhydride, benzoic acid, 4-methoxy-, anhydride, p-methoxybenzoic anhydride, 4-anisic anhydride, 4-methoxybenzoic acid anhydride, 4-methoxybenzoyl 4-methoxybenzoate, 4-methoxyphenyl anhydride, anisic anhydride
PubChem CID 69928
IUPAC Name (4-methoxybenzoyl) 4-methoxybenzoate
SMILES COC1=CC=C(C=C1)C(=O)OC(=O)C2=CC=C(C=C2)OC

Specifications

Melting Point 95°C to 99°C
Quantity 5 g
Beilstein 545712
Sensitivity Moisture sensitive
Solubility Information Soluble in alcohol,ether. Insoluble in water.
Formula Weight 286.29
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzoic anhydride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.