missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzenesulfonyl Chloride, TRC

Product Code. 30482861
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30482861 50 g 50g
30478082 5 g 5g
30467492 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482861 Supplier Toronto Research Chemicals Supplier No. TRCM33248550G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-68-0
Molecular Formula C7H7ClO3S
Molecular Weight (g/mol) 206.65
IUPAC Name 4-methoxybenzenesulfonyl chloride
SMILES COc1ccc(cc1)S(=O)(=O)Cl

Specifications

Quantity 50 g
InChI Formula InChI=1S/C7H7ClO3S/c1-11-6-2-4-7(5-3-6)12(8,9)10/h2-5H,1H3
Formula Weight 205.9804
Chemical Name or Material 4-Methoxybenzenesulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.