missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzenesulfonyl Chloride, 99%

Product Code. 10113370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10113370 100 g Glass bottle 100g
10501951 25 g Glass bottle 25g
10524274 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10113370 Supplier Thermo Scientific Acros Supplier No. 125731000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-68-0 | C7H7ClO3S | 206.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-68-0
Molecular Formula C7H7ClO3S
Molecular Weight (g/mol) 206.64
MDL Number MFCD00007446
InChI Key DTJVECUKADWGMO-UHFFFAOYSA-N
Synonym 4-methoxybenzene-1-sulfonyl chloride, p-anisylsulfonyl chloride, benzenesulfonyl chloride, 4-methoxy, 4-methoxybenzenesulphonyl chloride, 4-methoxybenzenesulfonylchloride, 4-methoxy-benzenesulfonyl chloride, p-anisolesulfonyl chloride, p-methoxyphenylsulfonyl chloride, 4-methoxyphenylsulfonyl chloride, benzenesulfonyl chloride, p-methoxy
PubChem CID 7401
IUPAC Name 4-methoxybenzenesulfonyl chloride
SMILES COC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 40.0°C to 42.0°C
Color Beige to White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4SO2Cl
Quantity 100 g
Beilstein 11, 243
Fieser 09,297
Solubility Information Solubility in water: decomposes
Formula Weight 206.65
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 4-Methoxybenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxybenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.