missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzenesulfonamide, 98%

Product Code. 11342806
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342806 5 g 5g
11352806 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342806 Supplier Thermo Scientific Alfa Aesar Supplier No. L08961.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1129-26-6 | C7H9NO3S | 187.213 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1129-26-6
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.213
MDL Number MFCD00025392
InChI Key MSFQEZBRFPAFEX-UHFFFAOYSA-N
Synonym 4-methoxybenzenesulphonamide, p-methoxybenzenesulfonamide, 4-methoxybenzene-1-sulfonamide, benzenesulfonamide, 4-methoxy, 4-methoxy-benzenesulfonamide, benzenesulfonamide, p-methoxy, benzenesulfonamide, p-methoxy-6ci,7ci,8ci, 3lka, p-anisolesulfonamide, acmc-1bvgf
PubChem CID 70789
IUPAC Name 4-methoxybenzenesulfonamide
SMILES COC1=CC=C(C=C1)S(=O)(=O)N

Specifications

Melting Point 110°C to 114°C
Quantity 5 g
Beilstein 2047881
Solubility Information Slightly soluble in water.
Formula Weight 187.22
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.