missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzamide, 98%

Product Code. 11475207
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475207 10 g 10g
11485207 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475207 Supplier Thermo Scientific Alfa Aesar Supplier No. B22614.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3424-93-9 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3424-93-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00007995
InChI Key GUCPYIYFQVTFSI-UHFFFAOYSA-N
Synonym p-anisamide, p-methoxybenzamide, benzamide, 4-methoxy, 4-methoxy benzamide, p-methyoxybenzamide, unii-z85fx3977e, 4-methoxy-benzamide, 4-anisamide, acmc-1cjjo, 4-methoxylbenzamide
PubChem CID 76959
IUPAC Name 4-methoxybenzamide
SMILES COC1=CC=C(C=C1)C(=O)N

Specifications

Melting Point 164°C to 167°C
Boiling Point 295°C
Quantity 10 g
Beilstein 1862847
Formula Weight 151.17
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.