missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzaldehyde dimethyl acetal, 98%

Product Code. 11432458
Change view
Click to view available options
Quantity:
25 g
250 g
500 g
Unit Size:
250g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432458 25 g 25g
11428700 250 g 250g
11442458 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11432458 Supplier Thermo Scientific Alfa Aesar Supplier No. A15793.14
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 2186-92-7 | C10H14O3 | 182.22 g/mol

4-Methoxybenzaldehyde dimethyl acetal is used as a precursor to 1-methoxy-1-(4-methoxyphenyl)hept-2-yne, 2,5-dioxopyrrolidin-1-yl-3-((4R)-2-(4-methoxyphenyl)-5,5-dimethyl-1,3-dioxane-4-carboxamido)propanoate and 1-(p-anisyl)-2-(2-methoxyethyl)-3-phenylindene. Further, it acts as a protecting group reagent for diols, especially in carbohydrates. It is also used as a flavor essence in sunflower, cyclamen and in jasmine. In addition to this, it is involved in allylation reactions with allyltrimethylsilane catalyzed by Iron(III) chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzaldehyde dimethyl acetal is used as a precursor to 1-methoxy-1-(4-methoxyphenyl)hept-2-yne, 2,5-dioxopyrrolidin-1-yl-3-((4R)-2-(4-methoxyphenyl)-5,5-dimethyl-1,3-dioxane-4-carboxamido)propanoate and 1-(p-anisyl)-2-(2-methoxyethyl)-3-phenylindene. Further, it acts as a protecting group reagent for diols, especially in carbohydrates. It is also used as a flavor essence in sunflower, cyclamen and in jasmine. In addition to this, it is involved in allylation reactions with allyltrimethylsilane catalyzed by Iron(III) chloride.

Solubility
Miscible with alcohol and many organic solvents. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2186-92-7
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00036507
InChI Key NNHYAHOTXLASEA-UHFFFAOYSA-N
Synonym 1-dimethoxymethyl-4-methoxybenzene, p-anisaldehyde dimethyl acetal, anisaldehyde dimethyl acetal, 4-methoxybenzaldehyde dimethyl acetal, p-dimethoxymethyl anisole, benzene, 1-dimethoxymethyl-4-methoxy, p-anisaldehydedimethylacetal, unii-xte77a5et4, anisicaldehyde dimethylacetal, p-anisaldehyde dimethylacetal
PubChem CID 75140
IUPAC Name 1-(dimethoxymethyl)-4-methoxybenzene
SMILES COC(OC)C1=CC=C(OC)C=C1

Specifications

Density 1.068
Boiling Point 253°C
Flash Point 97°C (206°F)
Odor Pleasant
Refractive Index 1.5055
Quantity 25 g
Beilstein 2048930
Solubility Information Miscible with alcohol and many organic solvents. Slightly miscible with water.
Formula Weight 182.22
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzaldehyde dimethyl acetal
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.