missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxyacetophenone, 98%

Product Code. 10091710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10091710 1 kg Glass bottle 1kg
10226010 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10091710 Supplier Thermo Scientific Acros Supplier No. 152360010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-06-1 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-06-1
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00008745
InChI Key NTPLXRHDUXRPNE-UHFFFAOYSA-N
Synonym 4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl
PubChem CID 7476
ChEBI CHEBI:86567
SMILES COC1=CC=C(C=C1)C(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 36.0°C to 39.0°C
Color White
Boiling Point 260.0°C
Flash Point 138°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4COCH3
Quantity 1 kg
Beilstein 08, 87
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
IUPAC Name 1-(4-methoxyphenyl)ethan-1-one
Formula Weight 150.18
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4'-Methoxyacetophenone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Methoxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.