missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-o-phenylenediamine dihydrochloride, 96%

Product Code. 11449503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449503 25 g 25g
11429503 1 g 1g
11439503 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449503 Supplier Thermo Scientific Alfa Aesar Supplier No. A12363.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 59548-39-9 | C7H12Cl2N2O | 211.086 g/mol

4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as derivatizing reagent by High-performance liquid chromatography (HPLC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-o-phenylenediamine is widely used as an intermediate in organic synthesis and pharmaceuticals. It is an important precursor for the synthesis of other heterocyclic compounds. It is used in the determination of glyoxal, methylglyoxal, and diacetyl in urine using 4-methoxy-o-phenylenediamine as derivatizing reagent by High-performance liquid chromatography (HPLC).

Solubility
Soluble in water.

Notes
Store in a cool place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59548-39-9
Molecular Formula C7H12Cl2N2O
Molecular Weight (g/mol) 211.086
MDL Number MFCD00052003
InChI Key SXCHMHOBHJOXGC-UHFFFAOYSA-N
Synonym 4-methoxybenzene-1,2-diamine dihydrochloride, 4-methoxy-o-phenylenediamine dihydrochloride, 3,4-diaminoanisole dihydrochloride, 1,2-benzenediamine, 4-methoxy-, dihydrochloride, 3,4-diaminoanisole 2hcl, 4-methoxy-phenylenediamine dihydrochloride, 4-methoxy-1,2-benzenediamine dihydrochloride, 4-methoxy-benzene-1,2-diamine dihydrochloride, 4-methoxy-1,2-phenylenediamine dihydrochloride, 4-methoxy-l,2-phenylene diamine dihydrochloride
PubChem CID 3085161
IUPAC Name 4-methoxybenzene-1,2-diamine;dihydrochloride
SMILES COC1=CC(=C(C=C1)N)N.Cl.Cl

Specifications

Melting Point ∼219°C (decomposition)
Quantity 25 g
Beilstein 3699636
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 211.09
Percent Purity ≥97%
Chemical Name or Material 4-Methoxy-o-phenylenediamine dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.