missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one, 97%, Thermo Scientific Chemicals

Product Code. 10246770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10246770 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10246770 Supplier Thermo Scientific Acros Supplier No. 235030050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82-57-5
MDL Number MFCD00005008
InChI Key NZVQLVGOZRELTG-UHFFFAOYSA-N
Synonym visnagin, desmethoxykhellin, visnagidin, visnagine, 5-methoxy-2-methylfuranochromone, 4-methoxy-7-methyl-5h-furo 3,2-g chromen-5-one, unii-p64438mlbw, 4-methoxy-7-methyl-furo 3,2-g chromen-5-one, chembl45176, 4-methoxy-7-methyl-5h-furo 3,2-g 1-benzopyran-5-one
PubChem CID 6716
ChEBI CHEBI:10002
IUPAC Name 4-methoxy-7-methylfuro[3,2-g]chromen-5-one
SMILES CC1=CC(=O)C2=C(O1)C=C3C(=C2OC)C=CO3

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 139°C to 142°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C13H10O4
Quantity 5 g
Merck Index 15, 10209
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in chloroform,sparingly soluble in alcohol
Molecular Weight (g/mol) 230.22
Formula Weight 230.22
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Methoxy-7-methyl-5H-furo[3, 2-g][1]benzopyran-5-one, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxy-7-methyl-5H-furo¢3,2-g!¢1!benzopyran-5-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.