missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-4-methyl-2-pentanone, 97%

Product Code. 11474527
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474527 100 g 100g
11464527 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474527 Supplier Thermo Scientific Alfa Aesar Supplier No. B21076.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-70-0 | C7H14O2 | 130.187 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-70-0
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00043932
InChI Key KOKPBCHLPVDQTK-UHFFFAOYSA-N
Synonym 4-methoxy-4-methyl-2-pentanone, pentoxone, pent-oxone solvent, 2-pentanone, 4-methoxy-4-methyl, 4-methyl-4-methoxy-2-pentanone, me 6 k, ccris 4840, acmc-1bwvk, dsstox_cid_5557, dsstox_rid_77828
PubChem CID 7884
IUPAC Name 4-methoxy-4-methylpentan-2-one
SMILES CC(=O)CC(C)(C)OC

Specifications

Boiling Point 156°C
Flash Point 48°C (118°F)
Odor Mild
Quantity 100 g
UN Number UN2293
Formula Weight 130.19
Percent Purity 97%
Chemical Name or Material 4-Methoxy-4-methyl-2-pentanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.