missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluoromethyl)aniline, 98+%, Thermo Scientific™

Product Code. 11410967
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11410967 10 g 10g
11400967 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410967 Supplier Thermo Scientific Alfa Aesar Supplier No. B23351.09

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 393-15-7
Molecular Formula C8H8F3NO
Molecular Weight (g/mol) 191.153
MDL Number MFCD00043460
InChI Key CQJCPOVTPNWVBW-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethyl aniline, 5-amino-2-methoxybenzotrifluoride, 3-trifluoromethyl-p-anisidine, 3-amino-6-methoxybenzotrifluoride, 4-methoxy-3-trifluoromethyl-phenylamine, benzenamine, 4-methoxy-3-trifluoromethyl, 4-methoxy-3-trifluoromethyl phenylamine, acmc-209j4q, 3-amino-6-methoxybenzotrifloride
PubChem CID 600705
IUPAC Name 4-methoxy-3-(trifluoromethyl)aniline
SMILES COC1=C(C=C(C=C1)N)C(F)(F)F

Specifications

Melting Point 57°C to 61°C
Boiling Point 112°C (18 mmHg)
Quantity 10 g
UN Number UN2811
Beilstein 2723972
Formula Weight 191.15
Percent Purity ≥98%
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.