missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-trifluoromethoxy-DL-phenylalanine, 97%, Thermo Scientific™

Product Code. 15438755
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1259979-50-4
Molecular Formula C11H12F3NO4
Molecular Weight (g/mol) 279.215
MDL Number MFCD09915503
InChI Key RAISOBRVUKCEEC-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethoxy-dl-phenylalanine, 2-amino-3-4-methoxy-3-trifluoromethoxy phenyl propanoic acid
PubChem CID 66523527
IUPAC Name 2-amino-3-[4-methoxy-3-(trifluoromethoxy)phenyl]propanoic acid
SMILES COC1=C(C=C(C=C1)CC(C(=O)O)N)OC(F)(F)F

Specifications

Formula Weight 279.21
Percent Purity 97%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material 4-Methoxy-3-trifluoromethoxy-DL-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'