missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-pyridineboronic acid hydrate, 97%

Product Code. 10711994 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10711994 1 g Glass bottle 1g
10609804 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10711994 Supplier Thermo Scientific Acros Supplier No. 378380010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1266615-79-5 | C6H8BNO3 | 152.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1266615-79-5 , 355004-67-0
Molecular Formula C6H8BNO3
Molecular Weight (g/mol) 152.94
MDL Number MFCD07186213
InChI Key YUTPAZKVEOJQCY-UHFFFAOYSA-N
Synonym 4-methoxy-3-pyridineboronic acid, 4-methoxypyridin-3-yl boronic acid, 4-methoxypyridine-3-boronic acid, 3-borono-4-methoxypyridine, 4-methoxy-3-pyridylboronic acid, boronic acid, 4-methoxy-3-pyridinyl, 4-methoxy-pyridine-3-boric acid, acmc-209iha, 4-methoxy-5-pyridineboronic acid, 4-methoxy-pyridin-3-boronic acid
PubChem CID 2762556
IUPAC Name (4-methoxypyridin-3-yl)boronic acid
SMILES COC1=CC=NC=C1B(O)O

Specifications

Color Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 152.94
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Methoxy-3-pyridineboronic acid hydrate
compliance-icons
Product Identifier
  • 4-Methoxy-3-pyridineboronic acid hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.