missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-nitrobenzonitrile, 97%, Thermo Scientific™

Product Code. 10612663
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 33224-23-6
Molecular Formula C8H6N2O3
Molecular Weight (g/mol) 178.15
MDL Number MFCD00221439
InChI Key ACAYRJJVKPMSIS-UHFFFAOYSA-N
PubChem CID 2800241
IUPAC Name 4-methoxy-3-nitrobenzonitrile
SMILES COC1=C(C=C(C=C1)C#N)[N+]([O-])=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-nitro-4-methoxybenzonitrile, 4-methoxy-3-nitro-benzonitrile, benzonitrile, 4-methoxy-3-nitro, 4-methoxy-3-nitrobenzenecarbonitrile, acmc-1cmtm, 5-cyano-2-methoxynitrobenzene, benzonitrile,4-methoxy-3-nitro
Formula Weight 178.15
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-nitrobenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'