missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-3-nitrobenzoic acid, 98%, Thermo Scientific™
£18.00 - £36.05
Chemical Identifiers
CAS | 89-41-8 |
---|---|
Molecular Formula | C8H7NO5 |
Molecular Weight (g/mol) | 197.15 |
MDL Number | MFCD00007256 |
InChI Key | ANXBDAFDZSXOPQ-UHFFFAOYSA-N |
Synonym | 4-methoxy-3-nitrobenzoic acid, 3-nitro-4-methoxybenzoic acid, 3-nitro-p-anisic acid, benzoic acid, 4-methoxy-3-nitro, unii-dd71o31olz, p-anisic acid, 3-nitro, dd71o31olz, 4-methoxy-3-nitro benzoic acid, 4-methoxy-3-nitro-benzoic acid, 4-methoxy-3-nitrobenzoicacid |
PubChem CID | 66640 |
SMILES | COC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10174010
|
Thermo Scientific Acros
187830250 |
25 g | Glass bottle |
£18.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10702811
|
Thermo Scientific Acros
187831000 |
100 g | Glass bottle | ||||||
Description
Chemical Identifiers
89-41-8 | |
197.15 | |
ANXBDAFDZSXOPQ-UHFFFAOYSA-N | |
66640 |
C8H7NO5 | |
MFCD00007256 | |
4-methoxy-3-nitrobenzoic acid, 3-nitro-4-methoxybenzoic acid, 3-nitro-p-anisic acid, benzoic acid, 4-methoxy-3-nitro, unii-dd71o31olz, p-anisic acid, 3-nitro, dd71o31olz, 4-methoxy-3-nitro benzoic acid, 4-methoxy-3-nitro-benzoic acid, 4-methoxy-3-nitrobenzoicacid | |
COC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-] |
Specifications
89-41-8 | |
100.0 | |
Yellow | |
98% | |
CH3OC6H3(NO2)CO2H | |
10, 181 | |
ANXBDAFDZSXOPQ-UHFFFAOYSA-N | |
197.15 | |
197.15 | |
Crystalline Powder |
97.5 | |
189°C to 194°C | |
Authentic | |
C8H7NO5 | |
MFCD00007256 | |
4-methoxy-3-nitrobenzoic acid, 3-nitro-4-methoxybenzoic acid, 3-nitro-p-anisic acid, benzoic acid, 4-methoxy-3-nitro, unii-dd71o31olz, p-anisic acid, 3-nitro, dd71o31olz, 4-methoxy-3-nitro benzoic acid, 4-methoxy-3-nitro-benzoic acid, 4-methoxy-3-nitrobenzoicacid | |
COC1=C(C=C(C=C1)C(=O)O)[N+](=O)[O-] | |
66640 | |
98% | |
4-Methoxy-3-nitrobenzoic acid, 98% |
Safety and Handling
missing translation for 'einecsNumber' : 201-906-0
missing translation for 'tsca' : TSCA
RUO – Research Use Only