missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-3-buten-2-one, tech. 90%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
4-Methoxy-3-buten-2-one is an important precursor for the synthesis of 1-methoxy-3-trimethylsilyloxy-1,3-butadiene and the Danishefsky diene and its equivalent. It is widely used as a reactant in the synthesis of hydroisoquinoline derivatives. It is employed as an intermediate for the synthesis of manzamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxy-3-buten-2-one is an important precursor for the synthesis of 1-methoxy-3-trimethylsilyloxy-1,3-butadiene and the Danishefsky diene and its equivalent. It is widely used as a reactant in the synthesis of hydroisoquinoline derivatives. It is employed as an intermediate for the synthesis of manzamine.
Solubility
Miscible with tetrahydrofuran, ether, acetonitrile and benzene.
Notes
Keep away from sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
4-Methoxy-3-buten-2-one is an important precursor for the synthesis of 1-methoxy-3-trimethylsilyloxy-1,3-butadiene and the Danishefsky diene and its equivalent. It is widely used as a reactant in the synthesis of hydroisoquinoline derivatives. It is employed as an intermediate for the synthesis of manzamine.
Solubility
Miscible with tetrahydrofuran, ether, acetonitrile and benzene.
Notes
Keep away from sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
Specifications
Density | 0.99 |
Boiling Point | 66°C to 69°C (12 mmHg) |
Flash Point | 63°C (145°F) |
Refractive Index | 1.465 |
Quantity | 250 g |
Beilstein | 1741053 |
Solubility Information | Miscible with tetrahydrofuran,ether,acetonitrile and benzene. |
Formula Weight | 100.12 |
Percent Purity | 90% |
Grade | Technical |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'