missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-buten-2-one, tech. 90%

Product Code. 11443564
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11443564 10 g 10g
11453564 50 g 50g
11463564 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11443564 Supplier Thermo Scientific Alfa Aesar Supplier No. A11235.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4652-27-1 | C5H8O2 | 100.12 g/mol

4-Methoxy-3-buten-2-one is an important precursor for the synthesis of 1-methoxy-3-trimethylsilyloxy-1,3-butadiene and the Danishefsky diene and its equivalent. It is widely used as a reactant in the synthesis of hydroisoquinoline derivatives. It is employed as an intermediate for the synthesis of manzamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-3-buten-2-one is an important precursor for the synthesis of 1-methoxy-3-trimethylsilyloxy-1,3-butadiene and the Danishefsky diene and its equivalent. It is widely used as a reactant in the synthesis of hydroisoquinoline derivatives. It is employed as an intermediate for the synthesis of manzamine.

Solubility
Miscible with tetrahydrofuran, ether, acetonitrile and benzene.

Notes
Keep away from sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4652-27-1
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008778,MFCD00008778,MFCD00008778
InChI Key VLLHEPHWWIDUSS-ARJAWSKDSA-N
Synonym z-4-methoxybut-3-en-2-one, 3z-4-methoxybut-3-en-2-one, 3-buten-2-one, 4-methoxy-, 3z, z-4-methoxy-3-butene-2-one
PubChem CID 643837
IUPAC Name (Z)-4-methoxybut-3-en-2-one
SMILES CO\C=C/C(C)=O

Specifications

Density 0.99
Boiling Point 66°C to 69°C (12 mmHg)
Flash Point 63°C (145°F)
Refractive Index 1.465
Quantity 10 g
Beilstein 1741053
Solubility Information Miscible with tetrahydrofuran,ether,acetonitrile and benzene.
Formula Weight 100.12
Percent Purity 90%
Grade Technical
Chemical Name or Material 4-Methoxy-3-buten-2-one
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.