missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-[[(RS)-(5-methoxy-1H-benzimidazol-2-yl)sulphinyl]methyl]-3,5-dimethylpyridine 1-Oxide (Omeprazole N-Oxide), Mikromol™

Product Code. 17983976
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17983976 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17983976 Supplier Mikromol Supplier No. MM0095.07
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Methoxy-2-[[(RS)-(5-methoxy-1H-benzimidazol-2-yl)sulphinyl]methyl]-3,5-dimethylpyridine 1-Oxide (Omeprazole N-Oxide)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Omeprazole N-Oxide
CAS 176219-04-8
Analyte or Component Names Omeprazole N-Oxide
Shipping Condition Room Temperature
Molecular Formula C17 H19 N3 O4 S
Certifications/Compliance ISO 9001
Synonym Esomeprazole Magnesium Trihydrate Imp. E (EP), Esomeprazole Imp. E (EP), Omeprazole Imp. E (EP), Omeprazole USP Related Compound E, Omeprazole USP RC E, 4-Methoxy-2-[[(RS)-(5-methoxy-1H-benzimidazol-2-yl)sulphinyl]methyl]-3,5-dimethylpyridine 1-Oxide, Omeprazole N-Oxide, Omeprazole Related Compound E, Esomeprazole Magnesium Trihydrate Impurity E, Esomeprazole Sodium Impurity E, Omeprazole Impurity E, Esomeprazole Impurity E, Esomeprazole Impurity E
InChI Formula InChI=1S/C17H19N3O4S/c1-10-8-20(21)15(11(2)16(10)24-4)9-25(22)17-18-13-6-5-12(23-3)7-14(13)19-17/h5-8H,9H2,1-4H3,(H,18,19)
SMILES COc1ccc2[nH]c(nc2c1)S(=O)Cc3c(C)c(OC)c(C)c[n+]3[O-]
IUPAC Name 5-methoxy-2-[(4-methoxy-3,5-dimethyl-1-oxidopyridin-1-ium-2-yl)methylsulfinyl]-1H-benzimidazole
Molecular Weight (g/mol) 361.42
Quantity 100 mg
Formula Weight 361.1096 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.